[(1R)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate

Lieferant: Apollo Scientific

PC450033-250MG
APOSPC450033-250MGEA 227 EUR
APOSPC450033-250MG
[(1R)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
[(1R)-1-(3,5-Bis(Trifluoromethyl)phenyl)ethyl][(1S)-1-phenylethyl]ammonium fumarate
Formel: C₁₈H₁₇F₆N·C₄H₄O₄
Molecular Weight: 477.397 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD25978114
CAS-Nummer: 672907-00-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR