6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Fluoro-1-isoindolinone, 6-Fluoroisoindolin-1-one
PC450035-1G
APOSPC450035-1GEA
139
EUR
APOSPC450035-1G
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
6-Fluoro-2,3-dihydro-1H-isoindol-1-one
Formel:
C₈H₆FNO Molecular Weight: 151.138 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09701283 CAS-Nummer: 340702-10-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...