4,6-Bis(trifluoromethyl)-2(1H)-pyrimidinethione

Lieferant: Apollo Scientific

PC450036-250MG
APOSPC450036-250MGEA 285 EUR
APOSPC450036-250MG
4,6-Bis(trifluoromethyl)-2(1H)-pyrimidinethione
4,6-Bis(trifluoromethyl)-2(1H)-pyrimidinethione
Formel: C₆H₂F₆N₂S
Molecular Weight: 248,149 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26408189
CAS-Nummer: 402520-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR