Bis(1H,1H-heptafluorobutyl)ether

Lieferant: Apollo Scientific

PC450043-1G
APOSPC450043-1GEA 720 EUR
APOSPC450043-1G
Bis(1H,1H-heptafluorobutyl)ether
Bis(1H,1H-heptafluorobutyl)ether
Formel: C₈H₄F₁₄O
Molecular Weight: 382,094 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123963
CAS-Nummer: 647-91-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR