Methyl (S)-2-fluoropropionate
Lieferant: Apollo Scientific
|
Achtung
|
PC450044-1G
APOSPC450044-1GEA
575
EUR
APOSPC450044-1G
Methyl (S)-2-fluoropropionate
Methyl (S)-2-fluoropropionate
Formel:
C₄H₇FO₂ Molecular Weight: 106,096 g/mol Siedepunkt: 35 °C (13 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD18072179 CAS-Nummer: 325690-13-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...