9,9-Bis[4-(1,2,2-trfluorovinyloxy)phenyl]-9H-fluorene

Lieferant: Apollo Scientific
Achtung

PC450045-250MG
APOSPC450045-250MGEA 80 EUR
APOSPC450045-250MG
9,9-Bis[4-(1,2,2-trfluorovinyloxy)phenyl]-9H-fluorene
9,9-Bis[4-(1,2,2-trfluorovinyloxy)phenyl]-9H-fluorene
Formel: C₂₉H₁₆F₆O₂
Molecular Weight: 510,427 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04038397
CAS-Nummer: 134130-20-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR