3-(Trifluoromethyl)phenyl fluorosulphonate
Lieferant: Apollo Scientific
PC450050-100MG
PC450050-250MG
APOSPC450050-100MGEA
109
EUR
APOSPC450050-100MG
APOSPC450050-250MG
3-(Trifluoromethyl)phenyl fluorosulphonate
3-(Trifluoromethyl)phenyl fluorosulphonate
Formel:
C₇H₄F₄O₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD24842695 CAS-Nummer: 1417220-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...