Tris(perfluorobutanesulphonyl)methane

Lieferant: Apollo Scientific
Gefahr

PC450052-1G
APOSPC450052-1GEA 314 EUR
APOSPC450052-1G
Tris(perfluorobutanesulphonyl)methane
Tris(perfluorobutanesulphonyl)methane
Formel: C₁₃HF₂₇O₆S₃
Molecular Weight: 862,295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489413
CAS-Nummer: 182505-69-7
UN: 3263
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR