4-Fluorophenyl(trifluoromethylsulphonyl)methane

Lieferant: Apollo Scientific

PC450062-1G
APOSPC450062-1GEA 548 EUR
APOSPC450062-1G
4-Fluorophenyl(trifluoromethylsulphonyl)methane
4-Fluorophenyl(trifluoromethylsulphonyl)methane
Formel: C₈H₆F₄O₂S
Molecular Weight: 242,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489418
CAS-Nummer: 2408-04-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR