2,6-Difluorophenyl(trifluoromethylsulphonyl)methane

Lieferant: Apollo Scientific

PC450066-1G
APOSPC450066-1GEA 328 EUR
APOSPC450066-1G
2,6-Difluorophenyl(trifluoromethylsulphonyl)methane
2,6-Difluorophenyl(trifluoromethylsulphonyl)methane
Formel: C₈H₅F₅O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD20489420
CAS-Nummer: 1346521-50-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR