2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
Lieferant: Apollo Scientific
|
Achtung
|
PC450067-1G
APOSPC450067-1GEA
967
EUR
APOSPC450067-1G
2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
2,2-Bis(1,3,5-trimethylphenyl)-1,1,1-trifluoroethane
Formel:
C₂₀H₂₃F₃ Schmelzpunkt: 115…119 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123955 CAS-Nummer: 1314534-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...