Bis(2,2,3,3-tetrafluoropropyl)ether

Lieferant: Apollo Scientific
Achtung

PC450069-250MG
APOSPC450069-250MGEA 137 EUR
APOSPC450069-250MG
Bis(2,2,3,3-tetrafluoropropyl)ether
Bis(2,2,3,3-tetrafluoropropyl)ether
Formel: C₆H₆F₈O
Molecular Weight: 246,098 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22123965
CAS-Nummer: 82914-35-0
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR