2-(Trifluoromethane)sulphonyl-1-[4-(trifluoromethoxy)phenyl]ethan-1-one

Lieferant: Apollo Scientific

PC450070-1G
APOSPC450070-1GEA 328 EUR
APOSPC450070-1G
2-(Trifluoromethane)sulphonyl-1-[4-(trifluoromethoxy)phenyl]ethan-1-one
2-(Trifluoromethane)sulphonyl-1-[4-(trifluoromethoxy)phenyl]ethan-1-one
Formel: C₁₀H₆F₆O₄S
Molecular Weight: 336,208 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489419
CAS-Nummer: 1346521-24-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR