1,1,1-Trifluoro-3-methylpentane

Lieferant: Apollo Scientific

PC450075-5G
APOSPC450075-5GEA 0 EUR
APOSPC450075-5G
1,1,1-Trifluoro-3-methylpentane
1,1,1-Trifluoro-3-methylpentane
Formel: C₆H₁₁F₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18074323
CAS-Nummer: 20733-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR