3-Bromo-2,3,3-trifluoropropene

Lieferant: Apollo Scientific

PC450080-1G
APOSPC450080-1GEA 236 EUR
APOSPC450080-1G
3-Bromo-2,3,3-trifluoropropene
3-Bromo-2,3,3-trifluoropropene
Formel: C₃H₂BrF₃
Molecular Weight: 174,947 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074337
CAS-Nummer: 220626-19-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR