Methyl perfluoropentyl ether

Lieferant: Apollo Scientific

PC450088-25G
APOSPC450088-25GEA 605 EUR
APOSPC450088-25G
Methyl perfluoropentyl ether
Methyl perfluoropentyl ether
Formel: C₆H₃F₁₁O
Lagertemperatur: Raumtemperatur
MDL: MFCD16620574
CAS-Nummer: 181214-74-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR