5-Bromo-4-chloro-4,5,5-trifluoropentanamide

Lieferant: Apollo Scientific

PC450092-5G
APOSPC450092-5GEA 458 EUR
APOSPC450092-5G
5-Bromo-4-chloro-4,5,5-trifluoropentanamide
5-Bromo-4-chloro-4,5,5-trifluoropentanamide
Formel: C₅H₆BrClF₃NO
Molecular Weight: 268.459 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074380
CAS-Nummer: 155630-24-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR