4-(2,2,3,3-Tetrafluoropropoxy)-1-nitrobenzene

Lieferant: Apollo Scientific

PC450093-5G
APOSPC450093-5GEA 575 EUR
APOSPC450093-5G
4-(2,2,3,3-Tetrafluoropropoxy)-1-nitrobenzene
4-(2,2,3,3-Tetrafluoropropoxy)-1-nitrobenzene
Formel: C₉H₇F₄NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00193786
CAS-Nummer: 109230-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR