(R)-1-(3,5-Difluorophenyl)ethanol

Lieferant: Apollo Scientific
Achtung

Synonyme: (R)-α-Methyl-3,5-difluorobenzyl alcohol

PC450096-1G
APOSPC450096-1GEA 430 EUR
APOSPC450096-1G
(R)-1-(3,5-Difluorophenyl)ethanol
(R)-1-(3,5-Difluorophenyl)ethanol
Formel: C₈H₈F₂O
Molecular Weight: 158,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18831076
CAS-Nummer: 433228-88-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR