(S)-1-(3,5-Difluorophenyl)ethylamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: (S)-1-(3,5-Difluorophenyl)ethanamine

PC450101-1G
APOSPC450101-1GEA 720 EUR
APOSPC450101-1G
(S)-1-(3,5-Difluorophenyl)ethylamine
(S)-1-(3,5-Difluorophenyl)ethylamine
Formel: C₈H₉F₂N
Molecular Weight: 157,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06761827
CAS-Nummer: 444643-16-7
UN: 2735
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR