2-chloro-5-iodo-3-(trifluoromethyl)pyridine

Lieferant: Apollo Scientific

PC450103-1G PC450103-250MG PC450103-5G
APOSPC450103-1GEA 0 EUR
APOSPC450103-1G APOSPC450103-250MG APOSPC450103-5G
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
2-chloro-5-iodo-3-(trifluoromethyl)pyridine
Formel: C₆H₂ClF₃IN
Lagertemperatur: Raumtemperatur
MDL: MFCD12024302
CAS-Nummer: 887707-25-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR