3,5-Difluoro-2,4,6-tris(2,2,3,3-tetrafluoropropoxy)pyridine
Lieferant: Apollo Scientific
PC450106-250MG
APOSPC450106-250MGEA
139
EUR
APOSPC450106-250MG
3,5-Difluoro-2,4,6-tris(2,2,3,3-tetrafluoropropoxy)pyridine
3,5-Difluoro-2,4,6-tris(2,2,3,3-tetrafluoropropoxy)pyridine
Formel:
C₁₄H₉F₁₄NO₃ Molecular Weight: 505,204 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19442155 CAS-Nummer: 1309602-69-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...