4-(2,2,3,3-Tetrafluoropropoxy)aniline

Lieferant: Apollo Scientific
Gefahr

PC450109-5G
APOSPC450109-5GEA 575 EUR
APOSPC450109-5G
4-(2,2,3,3-Tetrafluoropropoxy)aniline
4-(2,2,3,3-Tetrafluoropropoxy)aniline
Formel: C₉H₉F₄NO
Molecular Weight: 223,167 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00551780
CAS-Nummer: 58761-14-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR