(Perfluorohexyl)benzene
Lieferant: Apollo Scientific
PC450110-25G
APOSPC450110-25GEA
661
EUR
APOSPC450110-25G
(Perfluorohexyl)benzene
(Perfluorohexyl)benzene
Formel:
C₁₂H₅F₁₃ Molecular Weight: 396,147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042348 CAS-Nummer: 65440-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...