1,1-Difluoroethyl 2,2,3,3-tetrafluoropropyl ether
Lieferant: Apollo Scientific
PC450119-5G
APOSPC450119-5GEA
611
EUR
APOSPC450119-5G
1,1-Difluoroethyl 2,2,3,3-tetrafluoropropyl ether
1,1-Difluoroethyl 2,2,3,3-tetrafluoropropyl ether
Formel:
C₅H₆F₆O Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123966 CAS-Nummer: 1201897-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...