Perfluoro(1,2-diethylcyclohexane)

Lieferant: Apollo Scientific

PC450121-1G
APOSPC450121-1GEA 397 EUR
APOSPC450121-1G
Perfluoro(1,2-diethylcyclohexane)
Perfluoro(1,2-diethylcyclohexane)
Formel: C₁₀F₂₀
Lagertemperatur: Raumtemperatur
MDL: MFCD18072165
CAS-Nummer: 75169-49-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR