(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride

Lieferant: Apollo Scientific

Synonyme: (S)-1-(2-Fluoro-4-methoxyphenyl)ethanamine hydrochloride

PC450132-1G PC450132-250MG
APOSPC450132-1GEA 579 EUR
APOSPC450132-1G APOSPC450132-250MG
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride
(S)-1-(2-Fluoro-4-methoxyphenyl)ethylamine hydrochloride
Formel: C₉H₁₂FNO·ClH
Molecular Weight: 205.657 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16294167
CAS-Nummer: 1149383-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR