1-(Perfluorohexyl)docosane 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC450134-1G
PC450134-250MG
APOSPC450134-1GEA
408
EUR
APOSPC450134-1G
APOSPC450134-250MG
1-(Perfluorohexyl)docosane 95%
1-(Perfluorohexyl)docosane
Formel:
C₂₈H₄₅F₁₃ Schmelzpunkt: 53…55 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD28135666 CAS-Nummer: 825651-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...