[1H,1H-Perfluoro(2,5-dimethyl-3,6-dioxanonanoyl)] acrylate

Lieferant: Apollo Scientific

PC450143-5G
APOSPC450143-5GEA 458 EUR
APOSPC450143-5G
[1H,1H-Perfluoro(2,5-dimethyl-3,6-dioxanonanoyl)] acrylate
[1H,1H-Perfluoro(2,5-dimethyl-3,6-dioxanonanoyl)] acrylate
Formel: C₁₂H₅F₁₇O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD16621322
CAS-Nummer: 17559-01-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR