5-Fluoro-2-(2-pyridyl)-1H-benzimidazole

Lieferant: Apollo Scientific

PC450144-1G
APOSPC450144-1GEA 611 EUR
APOSPC450144-1G
5-Fluoro-2-(2-pyridyl)-1H-benzimidazole
5-Fluoro-2-(2-pyridyl)-1H-benzimidazole
Formel: C₁₂H₈FN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD12913332
CAS-Nummer: 875468-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR