3-(Trifluoromethoxy)benzaldehyde oxime

Lieferant: Apollo Scientific

PC450150-5G
APOSPC450150-5GEA 256 EUR
APOSPC450150-5G
3-(Trifluoromethoxy)benzaldehyde oxime
3-(Trifluoromethoxy)benzaldehyde oxime
Formel: C₈H₆F₃NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03410420
CAS-Nummer: 928063-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR