Perfluoro(4-methyl-2-pentenoic acid)
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(E)-Perfluoro(4-methylpent-2-enoic acid), Hexafluoro-4-(trifluoromethyl)pent-2-enoic acid, Nonafluoro(4-methylpent-2-enoic acid), 4-(Trifluoromethyl)hexafluoropent-2-enoic acid
PC450153-1G
APOSPC450153-1GEA
87.3
EUR
APOSPC450153-1G
Perfluoro(4-methyl-2-pentenoic acid)
Perfluoro(4-methyl-2-pentenoic acid)
Formel:
C₆HF₉O₂ Molecular Weight: 276.06 g/mol Schmelzpunkt: 173…174 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00153286 CAS-Nummer: 239795-58-5 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...