Trichloro(3,3,4,4,5,5,5-heptafluoropentyl)silane

Lieferant: Apollo Scientific

PC450155-1G
APOSPC450155-1GEA 783 EUR
APOSPC450155-1G
Trichloro(3,3,4,4,5,5,5-heptafluoropentyl)silane
Trichloro(3,3,4,4,5,5,5-heptafluoropentyl)silane
Formel: C₅H₄Cl₃F₇Si
Molecular Weight: 331,519 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22053870
CAS-Nummer: 866-24-0
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR