Methyl (R)-2-fluoropropionate
Lieferant: Apollo Scientific
|
Achtung
|
PC450159-1G
APOSPC450159-1GEA
1040
EUR
APOSPC450159-1G
Methyl (R)-2-fluoropropionate
Methyl (R)-2-fluoropropionate
Formel:
C₄H₇FO₂ Siedepunkt: 35 °C (13 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08457465 CAS-Nummer: 146805-74-5 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...