1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene

Lieferant: Apollo Scientific
Achtung

PC450160-1G
APOSPC450160-1GEA 85.7 EUR
APOSPC450160-1G
1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene
1-Bromo-4-(1,1,3,3,3-pentafluoropropoxy)benzene
Formel: C₉H₆BrF₅O
Molecular Weight: 305,039 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28054311
CAS-Nummer: 1823420-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR