3-(Trifluoromethyl)butyraldehyde 50% (w/w) in Toluol

Lieferant: Apollo Scientific

PC450162-5G
APOSPC450162-5GEA 183 EUR
APOSPC450162-5G
3-(Trifluoromethyl)butyraldehyde 50% (w/w) in Toluol
3-(Trifluoromethyl)butyraldehyde
Formel: C₅H₇F₃O
Molecular Weight: 140,104 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18074346
CAS-Nummer: 95853-69-3
UN: 1993
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR