5-Fluoro-2-(4-pyridyl)-1H-benzimidazole
Lieferant: Apollo Scientific
PC450167-1G
APOSPC450167-1GEA
195
EUR
APOSPC450167-1G
5-Fluoro-2-(4-pyridyl)-1H-benzimidazole
5-Fluoro-2-(4-pyridyl)-1H-benzimidazole
Formel:
C₁₂H₈FN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18074372 CAS-Nummer: 1309602-25-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...