Hexakis(1H,1H-perfluorobutoxy)phosphazene

Lieferant: Apollo Scientific
Achtung

PC450169-1G
APOSPC450169-1GEA 1210 EUR
APOSPC450169-1G
Hexakis(1H,1H-perfluorobutoxy)phosphazene
Hexakis(1H,1H-perfluorobutoxy)phosphazene
Formel: C₂₄H₁₂F₄₂N₃O₆P₃
Molecular Weight: 1329,223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22124072
CAS-Nummer: 470-73-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR