Hexakis(1H,1H-perfluorohexyloxy)phosphazene

Lieferant: Apollo Scientific
Achtung

PC450173-1G
APOSPC450173-1GEA 607 EUR
APOSPC450173-1G
Hexakis(1H,1H-perfluorohexyloxy)phosphazene
Hexakis(1H,1H-perfluorohexyloxy)phosphazene
Formel: C₃₆H₁₂F₆₆N₃O₆P₃
Lagertemperatur: Raumtemperatur
MDL: MFCD22124076
CAS-Nummer: 1365808-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR