Hexakis(1H,1H-nonafluoropentoxy)phosphazene 95%
Lieferant: Apollo Scientific
|
Achtung
|
PC450176-1G
APOSPC450176-1GEA
911
EUR
APOSPC450176-1G
Hexakis(1H,1H-nonafluoropentoxy)phosphazene 95%
Hexakis(1H,1H-nonafluoropentoxy)phosphazene
Formel:
C₃₀H₁₂F₅₄N₃O₆P₃ Molecular Weight: 1629,268 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22124073 CAS-Nummer: 1365808-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...