(1,2-Dibromo-2,2-difluoroethyl)benzene
Lieferant: Apollo Scientific
PC450183-1G
PC450183-250MG
PC450183-5G
APOSPC450183-1GEA
272
EUR
APOSPC450183-1G
APOSPC450183-250MG
APOSPC450183-5G
(1,2-Dibromo-2,2-difluoroethyl)benzene
(1,2-Dibromo-2,2-difluoroethyl)benzene
Formel:
C₈H₆Br₂F₂ Molecular Weight: 299.938 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134586 CAS-Nummer: 384-63-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...