Methyl undecafluoroamyl ketone
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl Perfluoropentyl Ketone, 1H,1H,1H-Undecafluoro-2-heptanone, 3,3,4,4,5,5,6,6,7,7,7-Undecafluoro-2-heptanone, Methyl Undecafluoropentyl Ketone, Methyl Perfluoroamyl Ketone
PC450188-5G
APOSPC450188-5GEA
265
EUR
APOSPC450188-5G
Methyl undecafluoroamyl ketone
Methyl undecafluoroamyl ketone
Formel:
C₇H₃F₁₁O Molecular Weight: 312,08 g/mol Dichte: 1,6 g/cm³ (25 °C) Flammpunkt: 32 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02093506 CAS-Nummer: 2708-07-8 UN: 1224 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...