1,1,1,4,4,4-Hexafluoro-2-(trifluoromethyl)-butane
Lieferant: Apollo Scientific
PC450191-1G
APOSPC450191-1GEA
575
EUR
APOSPC450191-1G
1,1,1,4,4,4-Hexafluoro-2-(trifluoromethyl)-butane
1,1,1,4,4,4-Hexafluoro-2-(trifluoromethyl)-butane
Formel:
C₅H₃F₉ Molecular Weight: 234,063 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD23134581 CAS-Nummer: 367-53-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...