3-Chloro-4-fluorobenzaldehyde oxime

Lieferant: Apollo Scientific
Achtung

PC450196-5G
APOSPC450196-5GEA 255 EUR
APOSPC450196-5G
3-Chloro-4-fluorobenzaldehyde oxime
3-Chloro-4-fluorobenzaldehyde oxime
Formel: C₇H₅ClFNO
Molecular Weight: 173,572 g/mol
Schmelzpunkt: 99…104 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00832721
CAS-Nummer: 253308-63-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR