3,5-Difluorobenzaldehyde oxime

Lieferant: Apollo Scientific
Achtung

PC450197-10G
APOSPC450197-10GEA 285 EUR
APOSPC450197-10G
3,5-Difluorobenzaldehyde oxime
3,5-Difluorobenzaldehyde oxime
Formel: C₇H₅F₂NO
Molecular Weight: 157,118 g/mol
Schmelzpunkt: 95…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03412421
CAS-Nummer: 677728-83-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR