2-(2,2-Difluoroethoxy)ethanol

Lieferant: Apollo Scientific
Achtung

PC450198-1G PC450198-25G PC450198-5G
APOSPC450198-1GEA 0 EUR
APOSPC450198-1G APOSPC450198-25G APOSPC450198-5G
2-(2,2-Difluoroethoxy)ethanol
2-(2,2-Difluoroethoxy)ethanol
Formel: C₄H₈F₂O₂
Lagertemperatur: Kühlschrank
MDL: MFCD19442036
CAS-Nummer: 853005-07-9
UN: 1993
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR