2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane

Lieferant: Apollo Scientific

PC450206-1G PC450206-5G
APOSPC450206-1GEA 116 EUR
APOSPC450206-1G APOSPC450206-5G
2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane
2-Iodo-2-(trifluoromethyl)-1,1,1,4,4,4-hexafluorobutane
Formel: C₅H₂F₉I
Lagertemperatur: Raumtemperatur
MDL: MFCD18074329
CAS-Nummer: 428-68-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR