1H,1H-Heptafluorobutyl butyrate
Lieferant: Apollo Scientific
PC450210-5G
APOSPC450210-5GEA
575
EUR
APOSPC450210-5G
1H,1H-Heptafluorobutyl butyrate
1H,1H-Heptafluorobutyl butyrate
Formel:
C₈H₉F₇O₂ Molecular Weight: 270,145 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22123991 CAS-Nummer: 2926-44-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...