2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol

Lieferant: Apollo Scientific
Achtung

PC450213-250MG PC450213-5G PC450213-1G
APOSPC450213-250MGEA 0 EUR
APOSPC450213-250MG APOSPC450213-5G APOSPC450213-1G
2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol
2-[2-(2,2-Difluoroethoxy)ethoxy]ethan-1-ol
Formel: C₆H₁₂F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD24842562
CAS-Nummer: 1435806-96-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR