2-(2,2,3,3-Tetrafluoropropoxy)ethanol

Lieferant: Apollo Scientific

PC450214-5G
APOSPC450214-5GEA 575 EUR
APOSPC450214-5G
2-(2,2,3,3-Tetrafluoropropoxy)ethanol
2-(2,2,3,3-Tetrafluoropropoxy)ethanol
Formel: C₅H₈F₄O₂
Molecular Weight: 176,109 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461945
CAS-Nummer: 50997-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR